Learn More
Uvex™ Oversleeves 9862 PE White 50 pair
Product Code. 17229911 Shop All Uvex Products
Click to view available options
Size:
Universal
Unit Size:
50 pairs
Product Code. 17229911
Brand: Uvex™ 9875300
Only null left
Add to basket
Add to basket
Gauntlets, Disposable chemical protection. Material: PP spunbond laminated with PE film
  • Gauntlets Disposable chemical protection Accessories
  • Color: white
  • Material: PP spunbond laminated with PE film
  • Elastic on both sides
  • 50 pairs in polybag
  • For single use only
  • Size: one size fits all
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Product Type Chemical-resistant Sleeve
Material Non-woven Microporous Polyethylene Laminate
Color White
Sterility Non-sterile
Style Standard Sleeve
Disposable Reusable
Seam Type Taped
Thickness 60 g/m²