Learn More
Honeywell Safety Products™ Overspec Safety Glasses Product Code.: 12697403
Honeywell Safety Products™ Overspec Safety Glasses
Product Code. 12697403
Lens Type: Impact Resistant
Click to view available options
Lens Type: Impact Resistant
Product Code. 12697403
Brand: Honeywell Safety Products™ 1027613
Quantity Price
1 £9.42 / Each
10 £8.45 / Each
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cover your prescription eyewear and guard your eyes from impact with these over-the-glass protective glasses. Honeywell Safety Products™ Overspec Safety Glasses offers all-day, comfortable coverage for a variety of workers and a wide range of prescription eyewear styles.
  • Over-the-glass coverage, offering workers full protection while wearing their own prescription eyewear.
  • Overspec offers all day coverage for a variety of workers and a wide range of prescription eyewear styles.
  • Bendable temples fit virtually all users.
  • Extremely comfortably and secure.
  • Temples curved outward to minimize temple interference with prescription eyewear .
  • Bendable temples to accommodate their face, head size and prescription frames.
  • Ratcheting temple (3 positions) to further customize the fit.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number C899.1B-D, EC Attestation Number C899.1B-D
Lens Coating Anti-fog, Impact-resistant
Product Type Safety glasses
For Use With (Application) Agriculture, Automotive, Building, Construction, Catering, Fishing, Food, Green Spaces, Laboratory, Logistics, Maintenance, Medical, Pharmaceutical, Service, Transportation and Wood industries.
Frame Color Blue, Black
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Nylon
Product Content Correction
Product Title
Honeywell Safety Products™ Overspec Safety Glasses >