missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ A1 Large Gel System
Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems, which have smaller footprints ideal for detailed RNA/DNA analysis.
Brand: Thermo Scientific™ A1-BP
Description
Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems. With one of the smallest footprints of any large-format device, these systems are ideal for detailed RNA/DNA analysis.
Features:
- Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
- Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
- Programmable power inverter may be used for field reversal electrophoresis
- UVT gel tray has gasketed end gates for leakproof seal without tape
- Built-in leveling ensures flat, uniformly thick gels
- Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
- Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
- Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Includes:
- Buffer chamber with three-point leveling base
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- UVT gel tray
- Gasketed end gates
- Three combs (12-, 16- and 20-well), 1.5mm thick
Specifications
CE marked | |
Owl | |
8 to 150 | |
14.8 x 8.5 x 4.3 in. (37.5 x 21.5 x 11 cm) | |
37.5 cm | |
8.5 in. | |
13 cm | |
11 cm |
Complete system with buffer exchange ports | |
Large, w/Buffer Exchange Ports | |
1.6 L | |
14.8 in. | |
25 cm | |
21.5 cm | |
4.3 in. |