missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ A2 and A5 Large Gel Electrophoresis System Standard Comb
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs, for use with Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems.
£48.30 - £49.50
Specifications
For Use With (Equipment) | Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems |
---|---|
Type | Comb |
Product Code | Brand | Description | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | |||
12413657
|
Fisherbrand™
A2-20C |
Standard comb | 20 | 1 mm |
£49.50
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
12413667
|
Fisherbrand™
A2-RL-18D |
Rapid Load comb | 18 | 1.5 mm |
£48.30
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||
Description
Eliminate comb assembly with an assortment of heavy-duty, one-piece Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems Combs, for use with Owl A2 and A5 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.Specifications
Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems | |
Comb |