missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ A2 Large Gel Systems

Product Code. 11520255
Click to view available options
Type:
Large Gel Electrophoresis System
Large, w/Buffer Exchange Ports
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11520255

Brand: Thermo Scientific™ A2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (16-, 24- and 36-well), 1.5mm thick
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Complete system
Product Line Owl
Type Large Gel Electrophoresis System
Sample Size 8 to 432 (16-, 24- and 36-well combs)
Volume (Metric) Buffer 2.3 L
Dimensions (L x W x H) 14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
Length (English) 14.8 in.
Length (Metric) 37.5 cm
Length (Metric) Gel 25 cm
Width (English) 12.8 in.
Width (Metric) 32.5 cm
Width (Metric) Gel 20 cm
Height (English) 4.1 in.
Height (Metric) 10.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.