missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ A5 Large Gel System with Built-In Recirculation
Get high resolution and uniform, reproducible results with these systems, featuring built-in recirculation. Ideal for long runs, multiple sample sets or RNA gels.
Brand: Thermo Scientific™ A5
This item is not returnable.
View return policy
Description
Get high resolution and uniform, reproducible results with Thermo Scientific™ Owl™ A5 Large Gel Systems. These systems feature built-in recirculation, and are ideal for long runs, multiple sample sets or RNA gels.
A5 Large Gel Systems:- Prevents the formation of pH and ionic gradients
- Provides high resolution and uniform, reproducible results
- Delivers clear results for samples run over long time periods
- Requires no external pumps, tubing or stir bars, because recirculation system is built into buffer chamber
- Eliminates uneven migration, band distortion or disassociation of pH-dependent glyoxylated RNA molecules that can result when ionic depletion occurs
Includes: Large gel system includes buffer chamber with three-point leveling base, SuperSafe™ lid with attached power supply leads, UVT gel tray with gasketed end gates, as well as 16-, 24-, and 36-well, 1.5m thick combs.
Specifications
CE marked | |
20 x 25 cm | |
Large Gel Electrophoresis System | |
2 L | |
14.8 in. | |
25 cm | |
32.5 cm | |
5.9 in. | |
250 V |
Gel Size: 20 x 25cm; Buffer volume: 2.0L; L x W x H: 37.5 × 32.5 × 15cm | |
Owl | |
8 to 432 | |
14.8 x 12.8 x 5.9 in. (37.5 x 32.5 x 15 cm) | |
37.5 cm | |
12.8 in. | |
20 cm | |
15 cm |