missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Aluminum-Backed Sequencers

Product Code. p-4528732
Click to view available options
Volume (Metric) Buffer:
800mL
950mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11913075

Brand: Thermo Scientific™ S3S

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Features:

  • Floating aluminum heat sink reduces chance of plates cracking during runs.
  • Upper buffer chamber (UBC) requires minimal buffer and is easily drained via inline drain port into removable bottle.
  • Lower buffer chamber (LBC) is removable for simple buffer disposal.
  • Side clamps press gel plates against gasket seated on UBC, applying even pressure across plate and forming a leak-proof seal resulting in flat, even banding patterns.
  • Clamps slide over glass plates for easy setup and removal of gels.

Includes:

  • Removable LBC
  • Heat sink
  • Two side clamps
  • Two blank glass plates
  • Two notched glass plates
  • Two sharktooth combs (78 teeth, 0.4mm thick)
  • Well comb (40 well, 0.4mm thick)
  • Two spacer sets (0.4mm)
  • Adhesive temperature indicator
  • UBC with removable drain bottle (with safety interlocking lid and attached power supply lead)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Gel: 35 x 45cm; 800mL; 23 x 44 x 50cm
Dimensions (L x W x H) Exterior 9 x 17.3 x 19.5 in. (44 x 23 x 50cm)
Format 40-well
Height (English) Exterior 19.5 in.
Height (Metric) Exterior 50 cm
Length (English) Exterior 9 in.
Length (English) Gel 17.716 in.
Length (Metric) Exterior 44 cm
Length (Metric) Gel 45 cm
Volume (English) Buffer 32.12 fl oz.
Volume (Metric) Buffer 950mL
Width (English) Exterior 17.3 in.
Width (English) Gel 13.77 in.
Width (Metric) Exterior 23 cm
Width (Metric) Gel 35 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.