missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System

Screen up to seven samples on one gel in less than five minutes with the Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System.

Brand:  Thermo Scientific™ C2-S

Product Code. 12434587

  • £254.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Screen up to seven samples on one gel in less than five minutes with the Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System.

Features:
  • Horizontal microgel system is designed for quick restriction fragment analysis.
  • Microgel may also be used to quickly check a few PCR samples before loading a large number of samples on a larger gel, saving time
  • Glass slides, 2 × 3 in. (5 × 7.5cm), serve as the gel tray
  • To use, pour and form thin gels by surface tension directly on glass slide; complete run lasts five to 10 minutes
  • Using available caster and casting slides, gels can be cast and solidified in less than 10 minutes.
Includes:
  • Buffer chamber (buffer volume, 100mL)
  • Lid
  • Power supply leads
  • Casting tray
  • 10 casting slides
  • One seven-well comb, 1.5mm thick
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Horizontal Electrophoresis System; Gel size: 5 x 7.5cm
Owl
7
6.75 x 2.63 x 4.38 in. (17 x 38 x 11 cm)
17 cm
2.63 in.
38 cm
4.38 in.
5 x 7.5 cm
Micro
100 mL
6.75 in.
7.5 cm
6.5
5 cm
11 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers