missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Casting Dams

Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.

Brand:  Thermo Scientific™ DAM-23

 View more versions of this product

Product Code. 12484617

  • £126.00 / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.

  • Seal agarose on contact, eliminating the need for tape
  • Does not attach to gel tray, enabling gels of any length to be poured
  • Easily handled and maneuvered using knob or finger depressions
  • Dam blocks off a portion of UVT gel tray
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Casting Dam, 23cm (L)
Owl
23 cm
Owl System
Gel Casting Dam
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers