Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.
£631.00 - £663.00
Specifications
Certifications/Compliance | CE marked |
---|---|
Gel Size | Mini |
Product Line | Owl |
Type | Mini Gel Electrophoresis System |
Sample Size | 5 to 34 |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11942935
|
Thermo Scientific™
B1-BP |
EasyCast w/buffer exchange ports |
£631.00
Each |
In Stock
Log in to see stock. |
|||||
11932935
|
Thermo Scientific™
B1 |
EasyCast |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.
Features:
- System offers a wide range of combs.
- Two comb slots on gel tray double sample capacity by doubling number of sample wells.
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
Includes:
- Main buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
- Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick
Ordering Information:
Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.
Warranty and Service Offering:
- Three years
Specifications
CE marked | |
Owl | |
5 to 34 | |
8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm) | |
22 cm | |
5.88 in. | |
9 cm | |
9.5 cm |
Mini | |
Mini Gel Electrophoresis System | |
600 mL | |
8.63 in. | |
11 cm | |
15 cm | |
3.75 in. |