missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel System Replacement Parts
Replace UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel System Replacement Parts, for use with the EasyCast™ B1 Mini Gel System.
Brand: Thermo Scientific™ B1-CST
This item is not returnable.
View return policy
Description
Replace your UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel System Replacement Parts, for use with the EasyCast™ B1 Mini Gel System.
For use with Owl EasyCast B1 Mini Gel Electrophoresis Systems.Specifications
External Caster (trays and combs not included) | |
Mini | |
Caster | |
11 cm |
Owl™ EasyCast B1 Mini Gel Electrophoresis System | |
Owl, EasyCast | |
36 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction