missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with these B1A Mini Gel Electrophoresis systems.
Brand: Thermo Scientific™ B1A
This item is not returnable.
View return policy
Description
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems. The ideal size for every lab, this system determines molecular weight ranges of PCR fragments or larger sized DNA molecules.
Features:- Two comb slots on gel tray double sample capacity by doubling number of sample wells.
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
- Leakproof casting allows a gel to be cast and run in the same chamber with no tape or additional parts required
- Run from five to 24 samples in 15 minutes.
Ordering Information:
Available with optional buffer exchange ports (B1A-BP) for use with an external recirculation pump.
Includes:
- Main buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Thermo Scientific™ EasyCast gasketed UVT gel tray
- Two heavy-duty, one-piece, double-sided combs: 6 & 10 well, 1.0/1.5mm thick
Specifications
CE marked | |
Mini | |
Mini Gel Electrophoresis System | |
400 mL | |
6.31 in. | |
8 cm | |
10.5 cm | |
3.75 in. | |
150 V |
EasyCast | |
Owl | |
5 to 24 | |
6.31 x 4.13 x 3.75 in. (16 x 10.5 x 9.5 cm) | |
16 cm | |
4.13 in. | |
7 cm | |
9.5 cm |