Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Brand:  Thermo Scientific™ B2-RL-25D
Code : 04
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12414577
  • £66.35 / Each
In Stock
to see stock.
Add to basket
Description
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8 and 12-channel pipette format (1X) and (2X) combs
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Specifications
Specifications
Rapid-load comb
Mini
Comb
1.5 mm
Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System
Owl, EasyCast
25 (2X)