Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Brand:  Thermo Scientific™ B2-RL-9C
Code : 04
Additional Details : Weight : 0.22000kg
 View more versions of this product
Product Code. 12424577
  • £34.10 / Each
Estimated Shipment: 07-10-2024
to see stock.
Add to basket
Description
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Ordering Information:
  • Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8- and 12-channel pipette format (1X) and (2X) combs
  • Customized combs are available (Order SKU#: XCM); please contact your customer service or sales representative for further details
Specifications
Specifications
Rapid-load comb
Mini
Comb
1 mm
Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System
Owl, EasyCast
9 (1X)