missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ Gel-Drying Supplies
Use the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
Brand: Thermo Scientific™ GDF-3
This item is not returnable.
View return policy
Description
Combine the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
- Frames and pre-cut cellophane available in large (24 × 24cm) or small (14 × 14cm) format
Specifications
Gel drying frame, Minigel | |
Gel Drying Frame | |
14 cm |
14 x 14 cm | |
Each | |
14 cm |