Learn More
Thermo Scientific™ Owl™ Gel Staining Box
Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method.
Brand: Thermo Scientific™ GSB-3
110.21 GBP valid until 2024-08-31
Use promo code "21655" to get your promotional price.
Code : 04
Additional Details : Weight : 0.43650kg
Description
Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method. Boxes hold up to 125mL of liquid and one gel when used on a shaking platform.
- Trays available in convenient size to hold the proper volume of staining reagents
- Grooved lid enables proper orientation and stability when stacking
- Made of clear plastic, so gels can be viewed directly in the tray
Ordering Information:
Package of five
Specifications
Gel Staining Box; Pack of 5 | |
125 mL | |
11 cm |
Pk. of 5 boxes | |
11 cm | |
3 cm |