missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ JGC Gel Casting Systems

Product Code. p-4528872 Shop All Thermo Scientific Products
Click to view available options
Length (Metric) Gel:
10 cm
14 cm
20 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11932975

Brand: Thermo Scientific™ JGC2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

  • Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
  • Design allows storage of gels without dehydration by adding buffer and heat sealing.
  • System casts from one to four gels.
  • Few moving parts result in outstanding durability, extra long life.

Includes:

  • Casting stand
  • Blocking plate
  • Three-point leveling base
  • Spacer placer
  • 25 gel pouches
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gel Electrophoresis
For Use With (Equipment) Owl JGC Gel Casting System
Gel Size 20 x 20 cm
Product Line Owl
Type Gel Caster
Length (Metric) Gel 14 cm
Width (English) 7.06 in.
Width (Metric) 18 cm
Width (Metric) Gel 16 cm
Depth (English) 6.69 in.
Depth (Metric) 17 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.