Learn More
Thermo Scientific™ Owl™ Leveling Platform
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
Brand:  Thermo Scientific™ B-LP
 View more versions of this product
Product Code. 12444557
  • £224.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
  • Size: 36 x 46cm
  • Includes bubble level
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Owl System
Owl
Leveling Platform