Learn More
Thermo Scientific™ P2-CST Multiple Gradient Gel Caster Comb, 25 teeth
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
Brand: Thermo Scientific™ P2-20FG
Code : 04
Additional Details : Weight : 2.21000kg
Description
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
- With an external gradient maker, gels can be cast with ease and great reproducibility
- The design and format of this unit will provide years of service in your lab
Specifications
Frosted blank glass | |
Blank Glass | |
20 cm |
Fisherbrand P2-CST System | |
3/16 in. | |
20 cm |