Learn More
Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster.
£31.20 - £210.00
Specifications
For Use With (Equipment) Owl P7-CST System
Product Line Owl
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12494687
View Documents
Thermo Scientific™
P7-10FG
Frosted blank glass
£31.20
Each
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
12404697
View Documents
Thermo Scientific™
P7-10FR
Frosted notched glass
£38.60
Each
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
12414697
View Documents
Thermo Scientific™
P7-CST
P7-CST Gradient Gel Caster
£210.00
Each
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
Description
Description
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster. One of the simplest devices to cast gradient gels, this caster is compatible with the Owl™ PBDS system.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
Specifications
Specifications
Owl P7-CST System
Owl