missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis Double Sided Vertical System, 8-10 x 10cm gel, 150-300mL buffer volume

Produce flat, even banding and crisp resolution with the easy-to-use Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis System.

Brand:  Thermo Scientific™ P82

Product Code. 12484697

  • £874.00 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Produce flat, even banding and crisp resolution with the easy-to-use Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis System.

Features:
  • Runs two 10 x 10cm gels or one gel when used with blocking plate; accommodates 8 x 10cm gels utilizing the provided adapter
  • Allows gels to easily be placed into the device
  • Wedge placed in front of the cassettes provides even pressure against the leakproof gasket, places the cassettes in proper running position
  • Two sizes of wedges accommodate varying thickness of precast gels
  • Flexible—use precast polyacrylamide gels or hand cast gels
  • No cooling required

Includes:

  • Inner buffer chambers
  • Buffer tank
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Two wedges (1.27cm thick)
  • Two wedges (1.59cm thick)
  • Adapter plates for 10 × 8 gels (2)
  • Blocking plate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE marked
15 mA
Dual-Gel Electrophoresis Double Sided Vertical System
150 mL
5.88 x 4.75 x 6.69 in. (15.2 x 12.4 x 17 cm)
15.2 cm
4.75 in.
10 cm
17 cm
Double Sided Vertical System, 8-10 x 10cm gel system, 150mL to 300mL buffer volume
Owl
300 mL
150 mL
5.88 in.
8 to 10 cm
12.4 cm
6.69 in.
330 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.