missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System.

Brand:  Thermo Scientific™ VEP-2

924.11 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 12494737

  • £1680.00 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System, ideal for fast, efficient transfer of proteins over a wide molecular weight range.

Features:
  • Tank-style electroblotting system
  • Integrated cooling base enables temperature-controlled runs
  • Stir bars (not included) may be added to bottom of buffer chamber for increased buffer circulation

Includes:

  • Buffer chamber with platinum wire electrode panels and cooling base
  • Safety interlocking lid with attached power supply leads
  • Four blotting cassettes with foam pads and cassette clips
  • Six sheets of blotting filter paper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VEP-2 mini Tank Electroblotting System, 9 x 9 in. gel size
Owl
0.3 gal.
7.06 x 5.13 x 6.5 in. (18 x 13 x 16.5 cm)
3.5 in.
9 cm
3.5 in.
9 cm
16.5 cm
Mini (9 in. x 9 in.)
Electroblotting System
1.4 L
7.06 in.
18 cm
5.13 in.
13 cm
6.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers