missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System

Rapidly transfer high molecular weight proteins with the Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System.

Brand:  Thermo Scientific™ VEP-3

Product Code. 12404747

  • £1340.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly transfer high molecular weight proteins with the Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System.

Features:
  • Removable cooling block permits efficient heat transfer.
  • Coded blotting cassettes ensure proper anode/cathode orientation; load easily into buffer chamber.
  • Stir bars (sold separately) may be added to bottom of buffer chamber to circulate buffer.
  • Cassettes allow easy loading of the gel sandwich and locks securely for loading into the buffer chamber.

Includes:

  • Buffer chamber with stainless-steel cathode plate and titanium anode plate
  • Safety interlocking lid with attached power supply leads and cooling module
  • Two blotting cassettes with foam pads
  • 25 sheets of 20W × 18.5cmL (7.88 × 7.25 in.) blotting filter paper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Large Tank Electroblotting System
Owl
1.05 gal.
3.5 x 11.8 x 30 in. (9 x 30 x 30 cm)
7.8 in.
20 cm
7.2 in.
18.5 cm
30 cm
20 x 18.5 cm
Electroblotting System
4 L
3.5 in.
9 cm
11.8 in.
30 cm
30 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers