missing translation for 'onlineSavingsMsg'
Learn More
Oxalic acid, 5% w/v aq. soln., Thermo Scientific Chemicals Product Code.: 11313737

Oxalic acid, 5% w/v aq. soln., Thermo Scientific Chemicals

Product Code. 11313737
1 L
Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11313737

Brand: Thermo Scientific Alfa Aesar 035619.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

UN Number UN3265
Concentration 5% w/v aq. soln.
Formula Weight 90.04
Quantity 1 L
Odor Odorless
Physical Form Liquid
Chemical Name or Material Oxalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM11L511 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog Number035619.K7,035619.K2
Lot NumberW26K511 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number035619.K7,035619.K2
Lot NumberN28K529 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number035619.K7,035619.K2
Lot NumberQ07J564 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog Number035619.K2
Lot NumberQ07J563 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog Number035619.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxalic acid, 5% w/v aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.