missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10523314
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Melting Point 98.0°C to 102.0°C
Assay Percent Range 99+%
Linear Formula HO2CCO2H·2H2O
Beilstein 02, 502
Fieser 01,764; 02,301; 05,481
Merck Index 15, 7010
Solubility Information Solubility in water: 138g/L (20°C). Other solubilities: 1200g/L in water (90°C), 400g/L in ethanol, 555g/L in boiling alcohol, 181g/L in glycerol, 10g/L in ether
Formula Weight 126.04
Percent Purity ≥99%
Grade Extra Pure
Quantity 5 kg
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Low Melting Crystalline Mass, Powder, Crystals and/or Chunks
Chemical Name or Material Oxalic acid dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'