missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, Extra Pure, SLR, Fisher Chemical™
£134.00 - £556.00
Chemical Identifiers
CAS | 6153-56-6 |
---|---|
Molecular Formula | C2H6O6 |
Molecular Weight (g/mol) | 126.064 |
MDL Number | 149102 |
InChI Key | GEVPUGOOGXGPIO-UHFFFAOYSA-N |
PubChem CID | 61373 |
IUPAC Name | oxalic acid;dihydrate |
SMILES | C(=O)(C(=O)O)O.O.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10224110
|
Fisher Chemical
O/0600/53 |
500 g | Plastic powder jar |
£134.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10182150
|
Fisher Chemical
O/0600/60 |
1 kg | Plastic powder jar |
£168.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10456880
|
Fisher Chemical
O/0600/62 |
2.5 kg | Plastic powder jar |
£340.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10713811
|
Fisher Chemical
O/0600/65 |
5 kg | Plastic keg |
£556.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Oxalic Acid Dihydrate, Extra Pure, SLR
Chemical Identifiers
6153-56-6 | |
126.064 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
oxalic acid;dihydrate |
C2H6O6 | |
149102 | |
61373 | |
C(=O)(C(=O)O)O.O.O |
Specifications
6153-56-6 | |
C2H6O6 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
oxalic acid;dihydrate | |
61373 | |
21.5mbar 50 | |
98°C |
White | |
149102 | |
C(=O)(C(=O)O)O.O.O | |
126.064 | |
126.04g/mol | |
1.3 | |
Powder/Solid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Oxalic acid dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title