missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 0.1 N standard solution

Product Code. 10331374 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10331374

Brand: Thermo Scientific Acros 124280025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

Color Colorless
Linear Formula HO2CCO2H
Beilstein 02, 502
Merck Index 15, 7010
Solubility Information Solubility in water: soluble in water.
Concentration 0.0998 to 0.1002N (20°C)
Formula Weight 90.04
Density 0.9900g/mL
Quantity 2.5 L
Packaging Plastic bottle
Physical Form Liquid
Specific Gravity 0.99
Chemical Name or Material Oxalic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.