Learn More
Chemical Identifiers
CAS | 79-37-8, 75-44-5, 76-02-8 |
---|---|
Molecular Formula | C2Cl2O2 |
Molecular Weight (g/mol) | 126.93 |
MDL Number | MFCD00000704 |
InChI Key | CTSLXHKWHWQRSH-UHFFFAOYSA-N |
Synonym | oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 |
PubChem CID | 65578 |
IUPAC Name | oxalyl dichloride |
SMILES | C(=O)(C(=O)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10113280
|
Thermo Scientific Acros
129610250 |
25 g | Glass bottle |
£34.67
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497900
|
Thermo Scientific Acros
129611000 |
100 g | Glass bottle |
£62.71
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10234990
|
Thermo Scientific Acros
129610010 |
1 kg | Glass bottle |
£309.54
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247183
|
Thermo Scientific Acros
129610025 |
2.5 kg | Glass bottle |
£703.50
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentChemical Identifiers
79-37-8, 75-44-5, 76-02-8 | |
126.93 | |
CTSLXHKWHWQRSH-UHFFFAOYSA-N | |
65578 | |
C(=O)(C(=O)Cl)Cl |
C2Cl2O2 | |
MFCD00000704 | |
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 | |
oxalyl dichloride |
Specifications
79-37-8, 75-44-5, 76-02-8 | |
Colorless to Yellow | |
63.0°C to 64.0°C (763.0 mmHg) | |
Authentic | |
C2Cl2O2 | |
1.4285 to 1.4305 | |
02, 542 | |
15, 7013 | |
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 | |
CTSLXHKWHWQRSH-UHFFFAOYSA-N | |
oxalyl dichloride | |
65578 | |
98% | |
Oxalyl chloride |
-12.0°C | |
1.4780g/mL | |
>100°C | |
97.5% min. (GC) | |
ClCOCOCl | |
MFCD00000704 | |
01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241 | |
1.478 | |
Solubility in water: reacts. Other solubilities: soluble in the main common organic solvents :, methylene chloride; dichloroethane; chlorobenzene;, ethylacetate; hexane; acetone; toluene; ether;, benzene; chloroform and ether | |
C(=O)(C(=O)Cl)Cl | |
126.93 | |
126.93 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Contact with w
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF INHA
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-200-2
missing translation for 'rtecsNumber' : KI2950000
missing translation for 'tsca' : TSCA
- Oxalyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.