missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxazole-5-carboxylic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽245°C (decomposition) |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 113.07 |
Percent Purity | ≥98% |
Chemical Name or Material | Oxazole-5-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'