missing translation for 'onlineSavingsMsg'
Learn More
Oxfendazole, Thermo Scientific™ Product Code.: 17515585

Oxfendazole, Thermo Scientific™

Product Code. 17515585
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 17515585

Brand: Thermo Scientific Acros 469210050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 53716-50-0
Molecular Formula C15H13N3O3S
Molecular Weight (g/mol) 315.35
MDL Number 00801063
InChI Key BEZZFPOZAYTVHN-UHFFFAOYNA-N
IUPAC Name methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1

Specifications

Packaging Glass Bottle
Quantity 5 g
Certificate of Analysis Available
Physical Form Powder/Crystals
Merck Index 13-7004
Color White to Light Yellow to Beige
Chemical Name or Material Oxfendazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxfendazole, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.