Learn More
WTW™ OxiCal -ST™ Calibration and storage vessel
The calibration and storage containers for the WTW oxygen sensors are suitable for the standard conform calibration (DIN ISO 5814 or DIN ISO 17289: 2014-12) with water steam saturated air as well as for storage of the sensors to protect the sensitive membrane.
Brand:  WTW™ 205349
Product Code. 10613782
  • £192.00 / Each
Estimated Shipment: 02-04-2025