Learn More
Diversey™ Oxivir Excel Concentrate
Highly concentrated one step cleaner disinfectant for non-invasive medical devices and nonporous surfaces.
Brand:  Diversey™ 100941436
164.95 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17231614
  • £192.00 / Pack of 2
Explore more special offers
Description
Description
  • Effective against Bacteria, Viruses and Yeasts
  • Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 sec
  • Extensively tested against European norms
  • Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
  • Excellent user safety profile reduces worker risk
  • No hazard warnings in dilution
  • Based on Diversey's patented AHP formulation (Accelerated Hydrogen Peroxide)
  • AHP formulation is gentle on surfaces extending equipment life
  • Does not contain respiratory irritants
  • Medical Device Class IIa
  • Oxivir Excel can be used on any hard surface usually encountered in a laboratory, production and non-production environments including stainless steel, glass, acrylics and PVC. It is not suitable for use on marble, copper, brass or other surfaces sensitive to oxidative agents.
Specifications
Specifications
Disinfectant Concentrate
Non-sterile
Jug
2 Pk.
1.95
Patented Accelerated Hydrogen Peroxide formulation (conc.)
For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only
5 L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Oxivir Excel
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Supplemental information
  • MIXTURE LIST-Contains : alkylbenzenesulphonic acid, propanol, hydrogen peroxide
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004