missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ Oxivir Excel Disinfectant Wipes
Pre-moistened cleaner disinfectant wipes for non-invasive medical devices and nonporous surfaces
Brand: Diversey™ 101104772
This item is not returnable.
View return policy
Description
- One-step cleaner disinfectant with broad spectrum efficacy and short contact times
- Oxivir Excel Wipe can be used on non-invasive medical devices and nonporous surfaces
- Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 seconds
- Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
- No hazard classification
- Powered by AHP Technology (Accelerated Hydrogen Peroxide)
- AHP formulation is gentle on surfaces extending equipment life
- Excellent user safety profile - no PPE required
- Does not contain respiratory irritants
- Suitable for use in high risk areas including ICU, OR, NICU and patient rooms
- Fragrance free
- No rinsing required
- Safer for the environment and user, since the AHP actives break down into water and oxygen. Free from Volatile Organic Compounds (VOCs)
- Medical Device Class IIa
Specifications
All Purpose Wipes | |
26 cm | |
20 cm | |
For BG and IE only | |
Flow Pack | |
100 Wipes/Pk., 24 Pk. | |
Efficacy proven by compliance with EN13727, EN13624, EN14476 and EN16615 |
10 in. | |
8 in. | |
Polypropylene | |
27 x 20 cm (10 x 8 in.) | |
All purpose virucidal wipes | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004
Spot an opportunity for improvement?Share a Content Correction