missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Agar Bacteriological

Product Code. 10055782
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
This item is not returnable. View return policy

Product Code. 10055782

Brand: Thermo Scientific™ LP0011T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.

Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.

TRUSTED_SUSTAINABILITY

Specifications

Description Agar No. 1, bacteriological
Form Powder
Product Type Agar
Quantity 5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.