missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Agar Technical

Click to view available options
Quantity:
5 kg
Unit Size:
5kg
Description
Oxoid Agar Technical (Agar No. 3) is a general-purpose bacteriological-grade agar.
- Normal working concentration 1.2%
Specifications
Specifications
| Description | Agar Tech (agar No 3) 5 Kg |
| Form | Powder |
| Product Type | Agar |
| Quantity | 5 kg |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction