missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ 2.5L Sachet
Thermo Scientific™ Oxoid AnaeroGen 2.5L Sachets are anaerobic gas generating sachets for use with Thermo Scientific™ Oxoid 2.5L jar, Part No. AG0025A (not included).
Brand: Thermo Scientific™ AN0025A
32.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Oxoid AnaeroGen 2.5L Sachets are anaerobic gas generating sachets for use with Thermo Scientific™ Oxoid 2.5L jar, Part No. AG0025A.
- No water or catalyst required
- No hydrogen gas produced

Specifications
Sachet | |
Thermo Scientific Oxoid | |
10/Pk. | |
2.5 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction