Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sachet
This compact sachet is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Brand: Thermo Scientific™ AN0020D
40.81 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sachet is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
- Use one sachet / Compact Plastic Pouch
- Use with Compact Plastic Pouch, Part No. AG0020C and AnaeroGen Compact Sealing Clip, Part No. AN0005C
Includes: 10 sachets.
Specifications
Compact Sachet | |
10 Sachets and 10 Pouches | |
10/Pk. |
Compact Gas Generation Sachet | |
Thermo Scientific Oxoid |