Learn More
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Brand: Thermo Scientific™ HP0053A
This item is not returnable.
View return policy
Description
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
For use only with Oxoid Antimicrobial Susceptibility Discs | |
Oxoid Antimicrobial Susceptibility Individual Disc Dispenser | |
Individual Disc Dispenser | |
6 Ejectors/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Antimicrobial Susceptibility Testing (AST) Range Brochure
File size: 2 MB
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility Individual disc Dispenser > 6/Pk
Spot an opportunity for improvement?Share a Content Correction