missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Basic Fuchsin Indicator

Oxoid Basic Fuchsin Indicator is a dye for use with Endo Agar Base, Part No. CM0479B and m Endo (LES) Agar Base, Part No. MM0551B.

Brand:  Thermo Scientific™ BR0050A

Product Code. 10401513

  • £35.65 / 10g
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Oxoid Basic Fuchsin Indicator is a dye for use with Endo Agar Base, Part No. CM0479B and m Endo (LES) Agar Base, Part No. MM0551B.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Basic Fuchsin Indicator
Powder
Indicator Dye
10 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Oxoid™ Basic Fuchsin Indicator > 10g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.