Learn More
Thermo Scientific™ Oxoid™ Bile Salts
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Brand: Thermo Scientific™ LP0055J
106.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Normal working concentration 0.5% w/v. Each vial supplements 500mL of medium.
Specifications
Bile Salts | |
Powder | |
Bile Salts | |
250 g |
Safety and Handling
- BILE SALTS L55
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium Taurocholate,Sodium Taurodeoxycholate, Sodium cholate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.