missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bismuth Sulphite Agar (Modified Wilson & Blair Medium) (Dehydrated)

Product Code. 11973382
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11973382

Brand: Thermo Scientific™ CM0201B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Salmonella typhi and other salmonllae from pathological material, sewage water supplies, food and other products with this medium.

Isolate Salmonella typhi and other salmonllae from pathological material, sewage water supplies, food and other products with Thermo Scientific™ Oxoid™ Bismuth Sulphite Agar (Modified Wilson & Blair Medium) (Dehydrated). Bismuth Sulphite Agar is a modification of the original Wilson and Blair1 Medium and is particularly useful for the isolation of lactose-fermenting salmonellae. The presence of bismuth sulphite and brilliant green make this medium highly selective for salmonellae.

Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.

Bismuth Sulphite Agar is a selective medium for the isolation and preliminary identification of Salmonella typhi and other salmonellae from pathological material, sewage, water supplies, food and other products suspected of containing these pathogens.

  • Selective: bismuth sulphite together with brilliant green acts as a selective agent making the medium selective for salmonellae while suppressing the growth of coliforms.
  • Easy-to-read: Salmonellae typically grow as black colonies with a surrounding metallic sheen resulting from hydrogen sulphide production and reduction of sulphite to black ferric sulphide.

Atypical colonies may appear if the medium is heavily inoculated with organic matter. Such a situation may be prevented by suspending the sample in sterile saline and using the supernatant for inoculation.

The freshly prepared medium has a strong inhibitory action2 and is suitable for heavily contaminated samples. Storing the poured plates at 4°C for 3 days causes the medium to change color to green, making it less selective with small numbers of salmonellae being recovered3. However, for Salmonella typhi recovery the latter technique is not recommended 4.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Maximum shelf life is 12 months.
Description Bismuth Sulphite Agar (Modified Wilson and Blair Medium)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 25.0L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.