missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Buffered Peptone Water
Brand: Thermo Scientific™ BO1067E
44.46 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.30000kg
Specifications
10 mL |