missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Sodium Chloride Peptone Solution EP/USP/JP/BP (Dehydrated)

Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP with this medium.

Brand:  Thermo Scientific™ CM0982B

93.35 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 10331053

  • £184.00 / 500g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP1,2,3 with Thermo Scientific™ Oxoid™ Buffered Sodium Chloride Peptone Solution (Dehydrated). The combination of phosphate buffer, sodium chloride and peptone increases the viability of sensitive microorganisms by providing an isotonic environment. This is of particular importance when trying to recover cells that may be stressed or sensitive to osmotic pressure.

Use Buffered Sodium Chloride Peptone Broth for the dilution of samples in the analysis of non-sterile products for microbial contaminants.

  • Conforms to the harmonized Microbial Limit Testing chapters from the EP/USP/BP/JP1,2,3
  • Maintains cell viability during the dilution process.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: EP.
Powder
500 g
Buffered Sodium Chloride Peptone Solution
Dehydrated Peptone
For 34.2L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only