Learn More
Thermo Scientific™ Oxoid™ CampyGen™ Compact Sachet
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen Compact Sachet.
Brand: Thermo Scientific™ CN0020C
74.65 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 50
Additional Details : Weight : 1.36080kg
Description
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen Compact Sachet.
- For use in Compact Plastic Pouch
- Each pouch holds 2 standard size petri dishes
- No water or catalyst required
- No hydrogen gas produced
- Use with Compact Plastic Pouch Part No. AG0020C and AnaeroGen™ Compact Sealing Clip, Part No. AN0005C
Specifications
CampyGen Compact Sachet | |
Compact Gas Generation Sachet | |
Thermo Scientific Oxoid | |
20/Pk. |