Learn More
Thermo Scientific™ Oxoid™ Clostridium difficile Test Kit
Thermo Scientific™ Oxoid Clostridium difficile Test Kit is a rapid latex agglutination test to identify C. difficile from culture media.
Brand: Thermo Scientific™ DR1107A
Code : 64
Additional Details : Weight : 0.16356kg
Description
Thermo Scientific™ Oxoid Clostridium difficile Test Kit is a rapid latex agglutination test used to identify Clostridium difficile from selective culture media.
Specifications
50 Tests/Kit |