Learn More
Thermo Scientific™ Reinforced Clostridial Medium (RCM) (Dehydrated)
Enumerate and cultivate clostridia and other anaerobes occurring in food and pathological samples with this medium.
Brand: Thermo Scientific™ CM0149B
117.78 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Cultivate and enumerate clostridia and other anaerobes occurring in food and pathological samples with Thermo Scientific™ Oxoid™ Reinforced Clostridial Medium. A semi-solid, highly nutritious medium can be used as an enrichment medium to optimize the growth of clostridia before sub-culturing to solid agar.
Use Reinforced Clostridial Medium (RCM) for the isolation and cultivation of anaerobic organisms occurring in a variety of habitats, including food and pathological specimens.
- Superior fertility: the medium produced the highest viable count when compared to other media tested by Hirsch & Grinstead 1
- Differential: can be made differential for sulphite-reducing clostridia by the addition of sodium sulphite and ferric citrate2
Reinforced Clostridial Medium (RCM) was designed by Hirsch & Grinstead1 for the cultivation and enumeration of clostridia. They showed that the medium was more fertile and enabled growth to be initiated from small inocula more readily than five other media tested. In a further comparison, the highest viable count obtainable was the criterion used, and again, RCM proved superior. Compared with the spleen infusion medium of Mundt & Jones3, RCM gave somewhat higher counts (Gibbs & Hirsch4).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
10°C to 30°C | |
Powder | |
Dehydrated Culture Media |
Reinforced Clostridial Medium (RCM) (Dehydrated) | |
Reinforced Clostridial Medium | |
500 g |