Learn More
Thermo Scientific™ Oxoid™ Diagnostic Discs
For identification and differentiation of microorganisms
Brand:  Thermo Scientific™ DD0002T
Code : 50
Additional Details : Weight : 0.01000kg
Product Code. 11973832
  • £10.55 / Pack of 50
In Stock
to see stock.
Add to basket
Description
Description
  • Differentiation of Lancefield Group A Streptococci from other beta-haemolytic Streptococci
  • Size: 50 discs
    • Order Info
      Supplied in cartridges of 50 discs each
Specifications
Specifications
Bacitracin
Product Suggestions
Product Suggestions
Videos
SDS Documents