Learn More
Thermo Scientific™ DrySpot™ E. coli O157 Latex Agglutination Test
Rapidly confirm E. coli serogroup O157 from colonies cultured on plated media by latex agglutination.
Brand: Thermo Scientific™ DR0120M
203.06 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 65
Additional Details : Weight : 0.01000kg
Description
The Oxoid DrySpot E. coli O157 test is a latex agglutination test for the identification of E. coli serogroup O157.
- Pre-dispensed reagents eliminate extra steps for faster time to result
- Simplified room temperature storage allows storage for up to two years
- Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
Specifications
2°C to 25°C | |
E. coli | |
40 reaction cards (3 tests/card), positive control, negative control, mixing paddles, bag clips |
DrySpot E. coli O157 Test Kit | |
Latex Agglutination Test | |
120 Tests/Kit |