missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DrySpot™ Pneumo Latex Agglutination Test

Rapidly detect capsular antigen from Streptococcus pneumoniae isolated from culture plates and blood culture by latex agglutination.

Brand:  Thermo Scientific™ DR0420M

Product Code. 11409878

  • £425.00 / 60 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Oxoid DrySpot™ Pneumo Test is a latex agglutination test for the detection of capsular antigen from Streptococcus pneumoniae to provide rapid identification of Strep. pneumoniae isolated from culture plates and blood culture.

Use this dry latex agglutination test for the identification of Streptococcus pneumoniae.

Blue latex particles sensitised with specific rabbit antibody are dried onto the test card. These react with recognised serological types of pneumococci to form agglutination and provide the test reaction area.

Blue latex particles sensitized with non-reactive rabbit globulins are dried onto the test card to provide the control reaction area.

Streptococcus pneumoniae
is a primary cause of bacterial pneumonia, meningitis and otitis media. The anti-phagocytic properties of the polysaccharide capsule are the key to the organism’s virulence. The organism may harmlessly inhabit the upper respiratory tract but may also gain access to the lungs by aspiration where it may establish an acute pneumonia. In addition, this organism also accesses the blood stream and the meninges to cause acute, purulent life-threatening infections.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 25°C
DrySpot Pneumo Latex Test
20 reaction cards (3 tests/card), positive control, negative control, phosphate buffered saline, mixing paddles, pouch clips
60 Tests/Kit
Positive
Streptococcus Test Kit
Latex
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6184788 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog NumberDR0420M
Lot Number6168114 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog NumberDR0420M
Lot Number6156743 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberDR0420M
Lot Number6142315 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberDR0420M
Lot Number6119219 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberDR0420M
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DrySpot™ Pneumo Latex Agglutination Test > 60 tests

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.