Learn More
Thermo Scientific™ Oxoid™ Egg Yolk Tellurite Emulsion
Brand: Thermo Scientific™ SR0054C
59.36 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 65
Additional Details : Weight : 0.20000kg
Description
A sterile emulsion of egg yolk containing potassium tellurite for use in Baird-Parker Medium (CM0275).Specifications
100 mL |